Alfa Aesar™ Glycidyl 2,2,3,3-tetrafluoropropyl ether, 97%

Manufacturer: Alfa Aesar™ H5350806

Quantity 5g

 View more versions of this product

Catalog No. AAH5350806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19932-26-4
    Molecular Formula C6H8O2F4
    Formula Weight 188.12
    MDL Number MFCD00054682
    Synonym 3-(2,2,3,3-Tetrafluoropropoxy)-1,2-propenoxide
    Chemical Name or Material Glycidyl 2,2,3,3-tetrafluoropropyl ether

    Specifications

    Formula Weight 188.12
    Molecular Formula C6H8O2F4
    MDL Number MFCD00054682
    CAS 19932-26-4
    Synonym 3-(2,2,3,3-Tetrafluoropropoxy)-1,2-propenoxide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Boiling Point 50°C (4mmHg)
    Quantity 5g
    Flash Point 79°C (174°F)
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Glycidyl 2,2,3,3-tetrafluoropropyl ether
    Assay 97%

    Description

    Applications
    Glycidyl 2,2,3,3-tetrafluoropropyl ether is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit