Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glycidyl phenyl ether, 99%

Click to view available options
Quantity:
50g
250g
1000g
122-60-1
C9H10O2
150.177
MFCD00005133
FQYUMYWMJTYZTK-UHFFFAOYSA-N
glycidyl phenyl ether,phenyl glycidyl ether,2-phenoxymethyl oxirane,1,2-epoxy-3-phenoxypropane,oxirane, phenoxymethyl,phenylglycidyl ether,2,3-epoxypropyl phenyl ether,phenoxypropene oxide,phenylglycydyl ether,ether, phenylglycidyl
31217
CHEBI:82367
2-(phenoxymethyl)oxirane
C1C(O1)COC2=CC=CC=C2
122-60-1
C9H10O2
150.177
MFCD00005133
FQYUMYWMJTYZTK-UHFFFAOYSA-N
glycidyl phenyl ether,phenyl glycidyl ether,2-phenoxymethyl oxirane,1,2-epoxy-3-phenoxypropane,oxirane, phenoxymethyl,phenylglycidyl ether,2,3-epoxypropyl phenyl ether,phenoxypropene oxide,phenylglycydyl ether,ether, phenylglycidyl
31217
CHEBI:82367
2-(phenoxymethyl)oxirane
C1C(O1)COC2=CC=CC=C2
Applications
2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

Solubility
Soluble in water (2.4g/L).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Flash Point 114°C (237°F)
Quantity 1000g
Odor Sweet
Melting Point 3.5°C
Boiling Point 244°C to 246°C
Beilstein 2744
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Glycidyl phenyl ether

GHS H Statement
H350-H341-H332-H315-H317-H335
May cause cancer.
Suspected of causing genetic defects.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : TZ3675000

TSCA : TSCA