Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl Phenyl Ether 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
122-60-1
C9H10O2
150.18
MFCD00005133
FQYUMYWMJTYZTK-UHFFFAOYSA-N
glycidyl phenyl ether,phenyl glycidyl ether,2-phenoxymethyl oxirane,1,2-epoxy-3-phenoxypropane,oxirane, phenoxymethyl,phenylglycidyl ether,2,3-epoxypropyl phenyl ether,phenoxypropene oxide,phenylglycydyl ether,ether, phenylglycidyl
31217
CHEBI:82367
2-(phenoxymethyl)oxirane
C1C(O1)COC2=CC=CC=C2
122-60-1
C9H10O2
150.18
MFCD00005133
FQYUMYWMJTYZTK-UHFFFAOYSA-N
glycidyl phenyl ether,phenyl glycidyl ether,2-phenoxymethyl oxirane,1,2-epoxy-3-phenoxypropane,oxirane, phenoxymethyl,phenylglycidyl ether,2,3-epoxypropyl phenyl ether,phenoxypropene oxide,phenylglycydyl ether,ether, phenylglycidyl
31217
CHEBI:82367
2-(phenoxymethyl)oxirane
C1C(O1)COC2=CC=CC=C2
Viscosity 7 mPa.s (20°C)
Specific Gravity 1.1
Melting Point 3.5°C
Refractive Index 1.5295 to 1.5315
Boiling Point 245.0°C
Quantity 50g
Packaging Glass bottle
Density 1.1000g/mL
Color Colorless to Yellow
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Solubility Information Solubility in water: 2.4g/L(20°C). Other solubilities: miscible with ethanol
Formula Weight 150.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Glycidyl phenyl ether, 99%
Show More Show Less

GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF ON SKIN: Wash with plenty of soap and water.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON

EINECSNumber : 204-557-2

RTECSNumber : TZ3675000

TSCA : TSCA