Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Glycine, 98.5 to 101.5% (Dry Basis), Thermo Scientific™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Plastic bottle
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Linear Formula H2NCH2CO2H
Packaging Plastic bottle
Formula Weight 75.07
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Melting Point ~245°C (decomposition)
Quantity 1kg
Specific Gravity 1.595
Chemical Name or Material Glycine
Assay Dry basis