Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number AC120070050
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O

Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Glycine is an essential acid that can be used for the synthesis of creatine, purines, and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity


Applications

  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
TRUSTED_SUSTAINABILITY
Melting Point 233°C
Arsenic (As) 3ppm max.
pH 5.5 to 6.5 (5% Aqueous Solution)
Color Colorless to Yellow
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 0.98
Linear Formula H2NCH2CO2H
Infrared Spectrum Authentic
Beilstein 04, 333
Fieser 1412
Loss on Drying 0.2% max. (105°C, 2 hrs)
Merck Index 15, 4526
Packaging Plastic Bucket
Residue after Ignition 0.1% max.
Formula Weight 75.07
Percent Purity ≥97.5% (Total Base)
Grade Extra Pure
Density 1.5950g/mL
Quantity 5 kg
Physical Form Liquid
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit