Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, 99%, ACS reagent, Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Assay Percent Range 99%
Linear Formula H2NCH2CO2H
Packaging Plastic bottle
Formula Weight 75.07
Grade ACS Reagent
Density 1.5950g/mL
Quantity 50g
Chemical Name or Material Glycine