Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Methyl Ester Hydrochloride 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
5680-79-5
C3H7NO2·HCl
125.56
MFCD00012870
COQRGFWWJBEXRC-UHFFFAOYSA-N
glycine methyl ester hydrochloride,h-gly-ome.hcl,methyl 2-aminoacetate hydrochloride,methyl glycinate hydrochloride,glycine, methyl ester, hydrochloride,methyl glycinate hcl,methyl aminoacetate hydrochloride,glycine methyl ester hcl,glycine methyl ester hydrochloride salt,aminoacetic acid methyl ester hydrochloride
122755
methyl 2-aminoacetate;hydrochloride
COC(=O)CN.Cl
5680-79-5
C3H7NO2·HCl
125.56
MFCD00012870
COQRGFWWJBEXRC-UHFFFAOYSA-N
glycine methyl ester hydrochloride,h-gly-ome.hcl,methyl 2-aminoacetate hydrochloride,methyl glycinate hydrochloride,glycine, methyl ester, hydrochloride,methyl glycinate hcl,methyl aminoacetate hydrochloride,glycine methyl ester hcl,glycine methyl ester hydrochloride salt,aminoacetic acid methyl ester hydrochloride
122755
methyl 2-aminoacetate;hydrochloride
COC(=O)CN.Cl
Packaging Plastic bottle
Melting Point 173°C to 177°C
Infrared Spectrum Authentic
Quantity 500g
Color White
Assay Percent Range 98%
Linear Formula H2NCH2CO2CH3·HCl
Beilstein 04,34
Solubility Information Solubility in water: >1000g/L water (20 c)
Formula Weight 125.56
Physical Form Powder or Cyrstals
Percent Purity 97.5 to 102.5% (Total base)
Chemical Name or Material Glycine methyl ester hydrochloride, 98%
Show More Show Less

EINECSNumber : 227-139-1