Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glycine, Molecular biology grade, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
500g
5kg
25kg
Packaging:
Fiber Carton
Plastic Barrel
Plastic Drum
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Packaging Plastic Barrel
Physical Form Crystalline powder
Percent Purity ≥98.5% by assay
Grade Molecular Biology Reagent
Color White
Quantity 25kg