Learn More
Promega Glycine, Molecular Biology Grade, Promega™
An amino acid used in the preparation of some electrophoresis buffers.
Manufacturer:  Promega H5071
 View more versions of this product
Catalog No. PRH5071
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Quality tested and certified free of DNase, RNase and protease activity
Specifications
Specifications
White
500 g
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
Crystalline
75.067
C2H5NO2
56-40-6
NCC(O)=O
75.07