Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glycine-N-methylamide hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
49755-94-4
C3H9ClN2O
124.568
MFCD00153440
ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
2-amino-n-methylacetamide hydrochloride,h-gly-nhme hcl,h-gly-nhme hydrochloride,acetamide, 2-amino-n-methyl-, monohydrochloride,h-gly-nhme.hcl,glycine methyl amide hcl,glycin-methylamid-hydrochlorid,2-amino-n-methylacetamide-hcl,n-methylglycinamide hydrochloride,glycine methyl amide hydrochloride
13842202
2-amino-N-methylacetamide;hydrochloride
CNC(=O)CN.Cl
49755-94-4
C3H9ClN2O
124.568
MFCD00153440
ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
2-amino-n-methylacetamide hydrochloride,h-gly-nhme hcl,h-gly-nhme hydrochloride,acetamide, 2-amino-n-methyl-, monohydrochloride,h-gly-nhme.hcl,glycine methyl amide hcl,glycin-methylamid-hydrochlorid,2-amino-n-methylacetamide-hcl,n-methylglycinamide hydrochloride,glycine methyl amide hydrochloride
13842202
2-amino-N-methylacetamide;hydrochloride
CNC(=O)CN.Cl
Linear Formula NH2CH2CONHCH3·HCl
Beilstein 3678660
Formula Weight 124.57
Physical Form Powder
Percent Purity 97%
Color White
Quantity 25g
Chemical Name or Material Glycine-N-methylamide hydrochloride

storageNote1 : 2° to 8°C