Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
56-40-6
C2H5NO2
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
NCC(O)=O
56-40-6
C2H5NO2
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
NCC(O)=O

AM125, 56-40-6, C2H5NO2

Spectrum™ Glycine, USP is a colorless, sweet-tasting crystalline non-essential amino acid. USP glycine has a wide variety of uses, including as an additive in pet food and animal feed, in foods and pharmaceuticals as a sweetener/taste enhancer, or as a component of food supplements and protein drinks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Chloride 7e-005
Ignition Residue 0.001
Loss on Drying 0.002
Packaging Fiber Drum
Percent Purity 98.5 to 101.5%
Sulfate 6.5e-005
Quantity 45 kg