Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Glycogen, from Oysters, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
5g
10g
25g
100g
Packaging:
Plastic bottle
9005-79-2
C24H42O21
666.579
MFCD00081547
BYSGBSNPRWKUQH-UJDJLXLFSA-N
glycogen,2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol,glycogen from bovine liver,2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol,2s,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol,4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439177
CHEBI:28087
(2S,3R,4S,5S,6R)-2-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O
9005-79-2
C24H42O21
666.579
MFCD00081547
BYSGBSNPRWKUQH-UJDJLXLFSA-N
glycogen,2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol,glycogen from bovine liver,2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol,2s,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol,4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439177
CHEBI:28087
(2S,3R,4S,5S,6R)-2-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O
Quantity 5g
Packaging Plastic bottle
Physical Form Powder
Grade Ultrapure
Chemical Name or Material Glycogen