Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycogen Synthase, p Ser641 Antibody (SR2258), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32152550UL
Description
Glycogen Synthase Monoclonal antibody specifically detects Glycogen Synthase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
Glycogen Synthase | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR2258 | |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
A synthesized peptide derived from human phospho-Glycogen synthase [Ser641] (Uniprot #: P13807) | |
50 μg | |
Phospho Specific | |
2997 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction