Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycol Sulfite 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3741-38-6
C2H4O3S
108.11
MFCD00005354
WDXYVJKNSMILOQ-UHFFFAOYSA-N
ethylene sulfite,glycol sulfite,1,3,2-dioxathiolane, 2-oxide,glycolsulfite,1,2-ethylene sulfite,cyclic ethylene sulfite,ethylene glycol cyclic sulfite,ethylene glycol, cyclic sulfite,ethylene sulphite,sulfurous acid, cyclic ester with ethylene glycol
77342
1,3,2-dioxathiolane 2-oxide
C1COS(=O)O1
3741-38-6
C2H4O3S
108.11
MFCD00005354
WDXYVJKNSMILOQ-UHFFFAOYSA-N
ethylene sulfite,glycol sulfite,1,3,2-dioxathiolane, 2-oxide,glycolsulfite,1,2-ethylene sulfite,cyclic ethylene sulfite,ethylene glycol cyclic sulfite,ethylene glycol, cyclic sulfite,ethylene sulphite,sulfurous acid, cyclic ester with ethylene glycol
77342
1,3,2-dioxathiolane 2-oxide
C1COS(=O)O1
Boiling Point 68°C (20.0mmHg)
Flash Point 79°C
Specific Gravity 1.42
Density 1.4200g/mL
Refractive Index 1.4450 to 1.447
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
Color Colorless
Assay Percent Range 98%
Formula Weight 108.11
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Glycol sulfite, 98%
Show More Show Less

EINECSNumber : 223-131-7

TSCA : TSCA