Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolic acid 70% in water, Thermo Scientific Chemicals

Catalog Number p-139196
Click to view available options
Quantity:
250 g
1 kg
25 kg
Packaging:
Glass bottle
Metal drum
79-14-1, 7732-18-5
C2H4O3
76.051
MFCD00004312
AEMRFAOFKBGASW-UHFFFAOYSA-N
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
757
CHEBI:17497
2-hydroxyacetic acid
C(C(=O)O)O
79-14-1, 7732-18-5
C2H4O3
76.051
MFCD00004312
AEMRFAOFKBGASW-UHFFFAOYSA-N
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
757
CHEBI:17497
2-hydroxyacetic acid
C(C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 10.0°C
Boiling Point 113.0°C
Color Yellow
Linear Formula CH2OHCOOH
Formula Weight 76.04
Percent Purity 70%
Density 1.2700 g/mL
Quantity 250 g
Packaging Glass bottle
Physical Form Solution
Specific Gravity 1.27
Chemical Name or Material Glycolic acid, 70% in water
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 201-180-5
RTECSNumber MC5250000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit