Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolic Acid, 70%, High Purity, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
79-14-1,7732-18-5
C2H4O3
76.05
AEMRFAOFKBGASW-UHFFFAOYSA-N
2-hydroxyacetic acid
OCC(O)=O
79-14-1,7732-18-5
C2H4O3
76.05
AEMRFAOFKBGASW-UHFFFAOYSA-N
2-hydroxyacetic acid
OCC(O)=O

G1059, 79-14-1, C2H4O3

Spectrum™ Glycolic Acid, 70 Percent, High Purity , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.
CAS Max % 0.7,0.3
UN Number UN3265
Heavy Metals (as Pb) 5e-006
Packaging Amber Glass Bottle
Water Approx. 30%
Quantity 4 x 4 L