Learn More
CD235a Mouse anti-Human, FITC, Clone: CLB-ery-1 (AME-1), Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHGLA014
 View more versions of this product
Catalog No. 50-112-1239
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients. This antibody recognizes a papain-sensitive epitope in the extracellular domain of the sialoglycoprotein known as glycophorin A (CD235a).
Glycophorin A is a sialoglycoprotein present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
CD235a
Monoclonal
FITC
GYPA
Liquid
GYPA
Human Glycophorin A (CD235a)
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CLB-ery-1 (AME-1)
PBS with BSA and 0.1% sodium azide
P02724
GYPA, glycophorin A, HGNC:4702, GPA, GPErik, GPSAT, HGpMiV, HGpMiX, HGpMiXI, HGpStaC, MN, MNS , Mi.V glycoprotein
Mouse
IgG1
2 mL
4° C, store in dark
Primary
2993