Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD235a Mouse anti-Human, FITC, Clone: CLB-ery-1 (AME-1), Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$240.00 - $808.00

Specifications

Antigen CD235a
Clone CLB-ery-1 (AME-1)
Target Species Human
Conjugate FITC
Isotype IgG1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-1239 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Invitrogen
MHGLA014
2 mL Each for $808.00
Add to cart
 
50-112-1238 Non-distribution product as customer accommodation. View Documents Invitrogen
MHGLA01
0.5 mL Each for $240.00
Add to cart
 
Description

Description

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients. This antibody recognizes a papain-sensitive epitope in the extracellular domain of the sialoglycoprotein known as glycophorin A (CD235a).

Glycophorin A is a sialoglycoprotein present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

CD235a
Human
IgG1
Monoclonal
Liquid
Antibody
4° C, store in dark
CLB-ery-1 (AME-1)
FITC
Flow Cytometry
Primary
Monoclonal
PBS with BSA and 0.1% sodium azide
ASR
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit