Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Antibody (MM0314-7S41) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$204.00 - $542.50
Specifications
Antigen | Glycophorin A |
---|---|
Clone | MM0314-7S41 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Glycophorin A | |
LYOPH | |
Unconjugated | |
Mouse | |
Cancer, Signal Transduction | |
AAA52768.1 | |
2993 | |
IgG1 | |
Protein G purified |
MM0314-7S41 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
GYPA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title