Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Mouse anti-Human, FITC, Clone: HIR2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143108F

Catalog No. NBP143108F


Add to cart

Description

Description

Glycophorin A Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glycophorin A
HIR2
Flow Cytometry
Purified
GYPA
The immunogen for this antibody was CD235a.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Glycophorin A antibody clone HIR2 recognizes N-terminal portion of glycophorin A (and weakly of glycophorin B). Its antigen is expressed on early erythroblasts, late erythroblasts, erythroblasts, mature erythrocytes and the cells of erythroid cell lines K562 and HEL, but not on all other cells.
Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Mouse
IgG2b Kappa
0.1 ml
Cancer, Signal Transduction
Primary
2993
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit