Learn More
Glycophorin A Mouse anti-Human, Phycoerythrin, Clone: HIR2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143108PE
Catalog No. NBP143108PE
Add to cart
Description
Description
Glycophorin A Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Glycophorin A
HIR2
Flow Cytometry
Purified
GYPA
The immunogen for this antibody was CD235a.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Glycophorin A antibody clone HIR2 recognizes N-terminal portion of glycophorin A (and weakly of glycophorin B). Its antigen is expressed on early erythroblasts, late erythroblasts, erythroblasts, mature erythrocytes and the cells of erythroid cell lines K562 and HEL, but not on all other cells.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Mouse
IgG2b Kappa
0.1 ml
Cancer, Signal Transduction
Primary
2993
Human