Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Mouse, Clone: SPM183, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Glycophorin A |
---|---|
Clone | SPM183 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Erythrocyte Agglutination, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, Agglutination, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247715B
|
Novus Biologicals
NBP2477150.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human, Bovine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Erythrocyte Agglutination, CyTOF-ready.Specifications
Glycophorin A | |
1.0 mg/mL | |
Flow Cytometry, Immunofluorescence, Agglutination, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Signal Transduction | |
PBS with No Preservative | |
2993 | |
Human erythrocytes treated with neuraminidase | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
39 kDa |
SPM183 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Erythrocyte Agglutination, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Bovine | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
GYPA | |
IgM κ | |
Protein A or G purified | |
Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A. It reacts with a peptide epitope on the extracellular domain of human glycophorin. Glycophorin A is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title