Learn More
Glycophorin C Mouse anti-Human, Alexa Fluor™ 532, Clone: BRIC4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233304AF532
Catalog No. NP233304A32
Add to cart
Description
Description
Glycophorin C Monoclonal antibody specifically detects Glycophorin C in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
Glycophorin C
BRIC4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2′, Sialoglycoprotein D
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
NBP2-33304 recognises the erythrocyte transmembrane glycoprotein CD236, also known as glycophorin C (GPC). CD236 is an integral membrane protein which carries the Gerbich blood group antigens. Expression of CD236 is not restricted to erythroid cells, but is broadly distributed in both erythroid and non-erythroid tissues. Clone BRIC4 specifically reacts with an epitope located within residues 2-21 of CD236.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
GYPC
Human erythrocytes
0.1 mL
Primary
2995
Human