Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxal sodium bisulfite hydrate, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 g
1 kg
Specifications
Melting Point | 193.0°C to 195.0°C |
Quantity | 1 kg |
Assay Percent Range | 97.5% min |
Linear Formula | [-CH(OH)SO3Na]2·xH2O |
Merck Index | 14,451 |
Solubility Information | Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol |
IUPAC Name | disodium 1,2-dihydroxyethane-1,2-disulfonate |
Formula Weight | 266.16 |
Percent Purity | 98% |
Infrared Spectrum | Authentic |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction