Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxal sodium bisulfite hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
332360-05-1
C2H4Na2O8S2·xH2O
266.16
JQYGKJGYGOFFIK-UHFFFAOYSA-L
glyoxal-sodium bisulfite monohydrate,glyoxal sodium bisulfite addition compound hydrate,1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate,1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt,c2h4o8s2.2na.h2o,glyoxal-sodium bisulfite monohydrate mi,glyoxal bis sodium hydrogen sulfite adduct hydrate,glyoxal sodium bisulfite addition compound hydrate,98,disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate,disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate
16211366
disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate
C(C(O)S(=O)(=O)[O-])(O)S(=O)(=O)[O-].O.[Na+].[Na+]
332360-05-1
C2H4Na2O8S2·xH2O
266.16
JQYGKJGYGOFFIK-UHFFFAOYSA-L
glyoxal-sodium bisulfite monohydrate,glyoxal sodium bisulfite addition compound hydrate,1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate,1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt,c2h4o8s2.2na.h2o,glyoxal-sodium bisulfite monohydrate mi,glyoxal bis sodium hydrogen sulfite adduct hydrate,glyoxal sodium bisulfite addition compound hydrate,98,disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate,disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate
16211366
disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate
C(C(O)S(=O)(=O)[O-])(O)S(=O)(=O)[O-].O.[Na+].[Na+]
Assay Percent Range 97.5% min
Linear Formula [-CH(OH)SO3Na]2·xH2O
Merck Index 14,451
Solubility Information Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 266.16
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 10% max. (120°C, 2 hrs)
Solubility Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol
Melting Point 193.0°C to 195.0°C
Quantity 25g
Chemical Name or Material Glyoxal sodium bisulfite hydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning