Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase I Mouse anti-Human, Clone: CPTC-GLO1-3, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Glyoxalase I |
---|---|
Clone | CPTC-GLO1-3 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122098
|
Novus Biologicals
NBP30731220UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Glyoxalase I | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
2739 | |
IgG1 | |
Protein A or G purified |
CPTC-GLO1-3 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
Recombinant full-length human Glyoxalase I protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title