Learn More
Glyoxalase I Mouse anti-Non-species specific, PerCP, Clone: 9F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259369PCP
Catalog No. NB259369PCP
Add to cart
Description
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
Specifications
Glyoxalase I
9F11
Flow Cytometry
Purified
GLO1
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary
2739
Nonspecies specific
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.