Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxalase I Mouse anti-Nonspecies specific, Allophycocyanin/Cy7, Clone: 9F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259369APCCY7

Catalog No. N259369APC7


Add to cart

Description

Description

Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Nonspecies specific samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glyoxalase I
9F11
Flow Cytometry
Conjugated, Purified
GLO1
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary
2739.0
Nonspecies specific
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit