Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxalbis(2-hydroxyanil), 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1149-16-2
C14H12N2O2
240.26
MFCD00002183
OMJNMRJMNUREOJ-KOBPNRPCSA-N
glyoxal bis 2-hydroxyanil,glyoxalbis 2-hydroxyanil,glyoxal bis o-hydroxyanil,glyoxal bis 2-hydroxyaniline,glyoxylidenebis 2-hydroxyaniline,unii-e2aw5xx09u,gbha,2,2'-ethanediylidenedinitrilo bisphenol,phenol, 2,2'-1,2-ethanediylidenedinitrilo bis,2,2'-ethanediylidenedinitrilo diphenol
5356911
6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one
C1=CC=C(C(=C1)NC=CN=C2C=CC=CC2=O)O
1149-16-2
C14H12N2O2
240.26
MFCD00002183
OMJNMRJMNUREOJ-KOBPNRPCSA-N
glyoxal bis 2-hydroxyanil,glyoxalbis 2-hydroxyanil,glyoxal bis o-hydroxyanil,glyoxal bis 2-hydroxyaniline,glyoxylidenebis 2-hydroxyaniline,unii-e2aw5xx09u,gbha,2,2'-ethanediylidenedinitrilo bisphenol,phenol, 2,2'-1,2-ethanediylidenedinitrilo bis,2,2'-ethanediylidenedinitrilo diphenol
5356911
6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one
C1=CC=C(C(=C1)NC=CN=C2C=CC=CC2=O)O
Indicator
Melting Point 201°C to 205°C
Infrared Spectrum Authentic
Sulfated Ash 0.1% max. (800°C)
Suitability suitable for determination of calcium
Packaging Glass bottle
Quantity 50g
Color Beige
Loss on Drying 0.5% max. (105°C)
Assay Percent Range 97%
Solubility Information Solubility in water: slightly soluble
Formula Weight 240.26
Physical Form Fine Powder
Percent Purity ≥96.0%
Chemical Name or Material Glyoxalbis(2-hydroxyanil)
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.