Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxylic acid monohydrate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
563-96-2
C2H2O3·H2O
92.06
MFCD00127974
MOOYVEVEDVVKGD-UHFFFAOYSA-N
glyoxylic acid monohydrate,glyoxylic acid hydrate,2-oxoacetic acid hydrate,oxaldehydic acid hydrate,glyoxalic acid monohydrate,glyoxalate, glyoxylate hydrate,glyoxalic acid hydrate,oxoacetate monohydrate,glyoxylic acid-hydrate,oxoacetic acid hydrate
15620607
oxaldehydic acid;hydrate
C(=O)C(=O)O.O
563-96-2
C2H2O3·H2O
92.06
MFCD00127974
MOOYVEVEDVVKGD-UHFFFAOYSA-N
glyoxylic acid monohydrate,glyoxylic acid hydrate,2-oxoacetic acid hydrate,oxaldehydic acid hydrate,glyoxalic acid monohydrate,glyoxalate, glyoxylate hydrate,glyoxalic acid hydrate,oxoacetate monohydrate,glyoxylic acid-hydrate,oxoacetic acid hydrate
15620607
oxaldehydic acid;hydrate
C(=O)C(=O)O.O
Infrared Spectrum Authentic
Melting Point 48.0°C to 52.0°C
Flash Point >110°C
Packaging Plastic bottle
Color White to Yellow
Quantity 25g
Assay Percent Range 98%
Linear Formula HCOCO2H·H2O
Solubility Information Solubility in water: freely soluble.
Formula Weight 92.06
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Grade Pure
Chemical Name or Material Glyoxylic acid monohydrate, 98%
Show More Show Less

GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti

GHS Signal Word: Danger