Learn More
Glypican 3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251751AP
Catalog No. 50-173-4769
Add to cart
Description
Description
GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predomitly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.
Specifications
Specifications
Glypican 3
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51654
DGSX, Glypican 3, GPC3, GTR2 2, Intestinal protein OCI 5, MXR7, OCI 5, OCI5, SDYS, SGB, SGBS, SGBS1
Rabbit
IgG
150 μL
-20°C
Primary
2719
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GPC3
Liquid
GPC3
Glypican 3 Fusion Protein Ag10129
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human