Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican 4 Mouse anti-Human, Alexa Fluor™ 647, Clone: 961609, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB9195R100UG

Catalog No. FAB9195R10


Add to cart
This item is not returnable. View return policy

Description

Description

Glypican 4 Monoclonal antibody specifically detects Glypican 4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glypican 4
961609
Flow Cytometry 0.25-1 μg/106 cells
O75487
glypican 4, glypican proteoglycan 4, glypican-4, GPC4, K-glypican, K-glypicandJ900E8.1 (glypican 4)
Chinese hamster ovary cell line CHO-derived recombinant human Glypican 4, Met1-Ser529, Accession # O75487
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Glypican 4 in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
2239
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit