Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Glypican 6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116163
Description
Glypican 6 Polyclonal Antibody for Western Blot, IHC (P), Flow
The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans. The glypicans have been implicated in the control of cell growth and division. Glypican 6 is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases.
Specifications
Glypican 6 | |
Polyclonal | |
Unconjugated | |
Gpc6 | |
glypican 6; glypican proteoglycan 6; glypican-6; GPC6; OMIMD1; Secreted glypican-6; UNQ369/PRO705 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
10082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1.8 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y625 | |
Gpc6 | |
KLH conjugated synthetic peptide between 496-528 amino acids from the C-terminal region of human GPC6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction