Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GlyT2/SLC6A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GlyT2/SLC6A5
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093973
View Documents
Novus Biologicals
NBP303334100UL
100 μg Each for $320.71
Add to cart
 
NB093974
View Documents
Novus Biologicals
NBP30333420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GlyT2/SLC6A5 Polyclonal antibody specifically detects GlyT2/SLC6A5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

GlyT2/SLC6A5
Unconjugated
Rabbit
Human, Mouse, Rat
GlyT2, GLYT-2, GLYT2GlyT-2, NET1, sodium- and chloride-dependent glycine transporter 2, solute carrier family 6 (neurotransmitter transporter, glycine), member 5, Solute carrier family 6 member 5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human GlyT2/SLC6A5 (NP_004202.3).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
9152
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit