Learn More
Alfa Aesar™ GM 6001
Manufacturer:  Alfa Aesar™ J65687LB0
 View more versions of this product
Catalog No. AAJ65687LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
GM 6001
142880-36-2
388.50
Moisture Sensitive
Off-white
Solid
A potent, cell-permeable, broad-spectrum hydroxamic acid inhibitor of matrix metalloproteinases
C20H28N4O4
Galardin; N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan methylamide