Learn More
Alfa Aesar™ GM 6001, Negative Control
Manufacturer:  Alfa Aesar™ J64386MCR
 View more versions of this product
Catalog No. AAJ64386MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1mg
-30°C to -10°C
Negative Control
C23H34N4O4
(N-t-Butoxycarbonyl-L-leucyl-L-tryptophan methylamide)
Soluble in DMSO
Off-white
Solid
A cell impermeant negative control for the MMP inhibitor GM 6001
430.54
Moisture Sensitive