Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Antibody (OTI8G5), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270462F

Catalog No. NBP270462F


Add to Cart

Description

Description

GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

GM-CSF
Monoclonal
FITC
PBS with 0.05% Sodium Azide
CSF2
Recombinant protein fragment corresponding to amino acids 18-144 of human GM-CSF (NP_000749) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Immunohistochemistry
OTI8G5
Immunohistochemistry
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Mouse
Affinity Purified
Biologically Active Proteins, Cytokine Research, Immunology
1437.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit