Learn More
Novus Biologicals™ GM-CSF Antibody Pair (HRP)
Solid phase sandwich ELISA for the quantitative determination of Human GM-CSF.
Manufacturer:  Novus Biologicals™ NBP2788445PLATES
 View more versions of this product
Catalog No. NB003006
Add to cart
Description
Description
Kit Components:
  • Capture antibody: Mouse anti-Human GM-CSF monoclonal antibody in PBS, pH 7.4, 1 mg/mL
  • Detection antibody: Mouse anti-Human GM-CSF monoclonal antibody conjugated to horseradish-peroxidase (HRP) in PBS, 50 % HRP-Protector, pH 7.4, 0.2 mg/mL
  • Standard: Highly purified HEK293-expressed recombinant Human GM-CSF protein, 35 ng
Specifications
Specifications
GM-CSF
HRP
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
5 Plates
Storage of components varies. See protocol for specific instructions.
1437
Human
ELISA
Sandwich ELISA
Mouse
Biologically Active Proteins, Cytokine Research, Immunology
Monoclonal
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human GM-CSF.
CSF2