Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Mouse anti-Human, Biotin, Clone: OTI8G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270462B

Catalog No. NBP270462B


Add to cart

Description

Description

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GM-CSF
OTI8G5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CSF2
Recombinant protein fragment corresponding to amino acids 18-144 of human GM-CSF (NP_000749) produced in E.coli.
Immunogen affinity purified
Biologically Active Proteins, Cytokine Research, Immunology
Primary
1437.0
Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit