Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA128021

Catalog No. PIPA128021


Add to Cart

Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GM-CSF Polyclonal antibody specifically detects GM-CSF in Rat samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GM-CSF
Polyclonal
Unconjugated
CSF2
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Goat
Antigen affinity chromatography
RUO
116630
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
0.1 mg/mL
PBS with no preservative
P48750
CSF2
Recombinant full length protein (Rat).
25 μg
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit