Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Human, Primate, Alexa Fluor™ 594, Clone: BVD2-21C11, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP234570AF594

Catalog No. NP23470A94


Add to cart

Description

Description

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human,Primate samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen).
Specifications

Specifications

GM-CSF
BVD2-21C11
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CSF2
Recombinant human GM-CSF protein
0.1 mL
Primary
1437
Human, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit