Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Mouse, PE, Clone: MP1-22E9, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 12733181

Encompass

 View more versions of this product

Catalog No. 50-111-96


Add to cart

Description

Description

Description: The MP1-22E9 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF). The MP1-22E9 antibody is a neutralizing antibody. Mouse GM-CSF is a 14 kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF-alpha, IL-2, IL-1, and IFN-gamma. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active. Applications Reported: MP1-22E9 has been reported for use in intracellular flow cytometric analysis. Applications Tested: This MP1-22E9 antibody is tested by intracellular staining and flow cytometric analysis of cultured mouse splencoytes. It is offered in 2 formats: - μg size: has been tested bu intracellular staining and flow cytometric analysis of cultured mouse splencoytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

- test size: has been pre-titrated and tested bu intracellular staining and flow cytometric analysis of cultured mouse splencoytes. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. GM-CSF (Granulocyte-Macrophage colony-stimulating factor) is a 14.6kDa hematopoietic growth factor that exists in glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The active form of the GM-CSF protein is found extracellularly as a homodimer and the GM-CSF gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q-syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. The ability of recombit GM-CSF to increase hematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation. Recent studies have investigated GM-CSF in inflammatory and autoimmune diseases such as arthritis, arthritis related interstitial lung disease, nephritis, and psoriasis. In the CNS, GM-CSF depletion or neutralization has been studied in experimental autoimmune encephalomyelitis (EAE) and multiple sclerosis (MS).
Specifications

Specifications

GM-CSF
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01587
Csf2
Affinity chromatography
RUO
12981
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MP1-22E9
PE
Csf2
Granulocyte Macrophage Colony Stimulating Factor, Colony-stimulating factor, CSF, Sargramostim, Molgramostin
Rat
50 μg
Primary
Mouse
Antibody
IgG2a κ
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.