Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130/GOLGA2 Rabbit anti-Human, Clone: 2059C, R&D Systems™

Rabbit Monoclonal Antibody

$125.35 - $329.00

Specifications

Antigen GM130/GOLGA2
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Format Purified
Clone 2059C
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB81991SP View Documents R&D Systems
MAB81991SP
25 ug Each for $125.35
Add to cart
 
MAB81991100 View Documents R&D Systems
MAB81991100
100 ug Each for $329.00
Add to cart
 
Description

Description

GM130, also known as Golgin subfamily A member 2 and GOLGA2, is a 130kDa member of the Golgin protein family. GM130/GOLGA2 is a component of the Golgi membrane involved in maintaining cis-Golgi structure. GM130/GOLGA2 protein is thought to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been predicted. Human GM130/GOLGA2 protein is 1002 amino acids (aa) in length, and over aa 528‑606, shares 90% sequence identity with its mouse and rat orthologs.
Specifications

Specifications

GM130/GOLGA2
Purified
Western Blot
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
2059C
Unconjugated
IgG
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit