Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GM130/GOLGA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF8199SP
This item is not returnable.
View return policy
Description
GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
GM130/GOLGA2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, GOLGA2, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
Sheep | |
Affinity Purified | |
RUO | |
2801 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 μg/mL, Simple Western 50 μg/mL, Immunocytochemistry 5-15 μg/mL | |
Q08379 | |
GOLGA2 | |
E. coli-derived recombinant human GM130/GOLGA2 Met528-Gln606 Accession # Q08379 | |
25 μg | |
Primary | |
Detects human GM130/GOLGA2 in direct ELISAs and Western blots. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction