Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM2 ganglioside Mouse anti-Human,Bovine, Unconjugated, Clone: GMB28, MilliporeSigma™

Mouse Monoclonal Antibody

$150.30 - $372.00

Specifications

Antigen GM2 ganglioside
Applications ELISA, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN25501MI
View Documents
MilliporeSigma
MABN2550100UL
100 μL
Each for $372.00
Add to cart
 
MABN25502MI
View Documents
MilliporeSigma
MABN255025UL
25 μL
Each for $150.30
Add to cart
 
Description

Description

GM2 ganglioside Monoclonal antibody specifically detects GM2 ganglioside in Human,Bovine samples. It is validated for ELISA, Immunohistochemistry (Paraffin).
Specifications

Specifications

GM2 ganglioside
Unconjugated
Mouse
Ganglioside GM2;GM2
Monoclonal
Ligatrap
ELISA, Immunohistochemistry (Paraffin)
Purified
Human, Bovine
Purified GM2 ganglioside attached to Salmonella minnesota.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit