Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gm5581 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191491

 View more versions of this product

Catalog No. NBP191491


Add to cart

Description

Description

Gm5581 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

Gm5581
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
EG434094, predicted gene 5581
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
434094
Mouse
Western Blot
Western Blot 1:1000
Affinity Purified
GM5581
The specific Immunogen is proprietary information. Peptide sequence EEWECLDSAQRDLYRDVMLENYHNLVSVGVAVSKSEVIFCLEQNNESWIA.
34 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit