Learn More
Bachem 4-Maleimidobutyric acid N-hydroxysuccinimide ester, 0.5g, 80307-12-6, MFCD00036817 DFS Item
Manufacturer:  Bachem 4087894.0500
GMBS (N-succinimidyl 4-maleimidobutyrate) is an often used heterobifunctional crosslinker, a standard reagent for conjugation of cysteine-containing peptides to carriers.
Catalog No. 50-194-7394