Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMCL1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549505

Catalog No. PA549505

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a nuclear envelope protein that appears to be involved in spermatogenesis, either directly or by influencing genes that play a more direct role in the process. This multi-exon locus is the homolog of the mouse and drosophila germ cell-less gene but the human genome also contains a single-exon locus on chromosome 5 that contains an open reading frame capable of encoding a highly-related protein.
Specifications

Specifications

GMCL1
Polyclonal
PBS with 0.09% sodium azide
Q96IK5, Q920G9
Germ cell-less protein-like 1, GMCL1, GCL
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23885, 64395
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GMCL1
Liquid
GMCL1
KLH conjugated synthetic peptide between 281-315 amino acids from the Central region of human GMCL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit