Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMDS Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2A1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270835AF647

Catalog No. N270835A647


Add to cart

Description

Description

GMDS Monoclonal antibody specifically detects GMDS in Human samples. It is validated for Western Blot.
Specifications

Specifications

GMDS
OTI2A1
Western Blot
Affinity Purified, Conjugated
GMDS
Full length recombinant protein of human GMDS (NP_001491) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
2762.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 4.2.1.47, GDP-D-mannose dehydratase, GDP-mannose 4,6-dehydratase, GMDGDP-mannose 4,6 dehydratase, SDR3E1, short chain dehydrogenase/reductase family 3E, member 1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit