Learn More
GMF-beta Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0317-8X12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212343AF750
Catalog No. NP212343A70
Add to cart
Description
Description
GMF-beta Monoclonal antibody specifically detects GMF-beta in Human samples. It is validated for Western Blot.
Specifications
Specifications
GMF-beta
MM0317-8X12
Western Blot
glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
GMFB
Human GMF beta
0.1 mL
Primary
2764